1,4-Bis[(9S)-10,11-dihydro-6′-methoxycinchonan-9-yl]-9,10-anthracenedione 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD134154-250MGEA 1380 DKK
BLDPBD134154-250MG BLDPBD134154-1G BLDPBD134154-100MG BLDPBD134154-5G
1,4-Bis[(9S)-10,11-dihydro-6′-methoxycinchonan-9-yl]-9,10-anthracenedione 98%
1,4-Bis[(9S)-10,11-dihydro-6′-methoxycinchonan-9-yl]-9,10-anthracenedione

Storage condition: Inert atmosphere, Room Temperature

Formel: C₅₄H₅₆N₄O₆
Molekylvægt: 857,0457 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD29075644
CAS-nummer: 176298-44-5

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR