(Z)-2-(2-Aminothiazol-4-yl)-2-(tert-Butoxycarbonylmethoxyimino)acetic acid 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD13324-250MGEA 46.6 DKK
BLDPBD13324-250MG BLDPBD13324-1G BLDPBD13324-5G
(Z)-2-(2-Aminothiazol-4-yl)-2-(tert-Butoxycarbonylmethoxyimino)acetic acid 98%
(Z)-2-(2-Aminothiazol-4-yl)-2-(tert-Butoxycarbonylmethoxyimino)acetic acid

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formel: C₁₁H₁₅N₃O₅S
Molekylvægt: 301,32 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD06411064
CAS-nummer: 74440-02-1

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR