Pyrrolidine-2-carboxamide 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD132900-5GEA 5400 DKK
BLDPBD132900-5G BLDPBD132900-250MG BLDPBD132900-1G BLDPBD132900-25G
Pyrrolidine-2-carboxamide 98%
Pyrrolidine-2-carboxamide

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₅H₁₀N₂O
Molekylvægt: 114,14 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD03788250
CAS-nummer: 2812-47-7

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR