(S)-Ethyl 2-amino-3-(4-chlorophenyl)propanoate hydrochloride 95+%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD13230-250MGEA 132 DKK
BLDPBD13230-250MG BLDPBD13230-1G BLDPBD13230-5G
(S)-Ethyl 2-amino-3-(4-chlorophenyl)propanoate hydrochloride 95+%
(S)-Ethyl 2-amino-3-(4-chlorophenyl)propanoate hydrochloride

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₁₁H₁₅Cl₂NO₂
Molekylvægt: 264,14 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00012531
CAS-nummer: 51366-22-4

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR