(S)-Methyl 1,2,3,4-tetrahydroisoquinoline-3-carboxylate hydrochloride 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD13228-250MGEA 202 DKK
BLDPBD13228-250MG BLDPBD13228-1G BLDPBD13228-5G
(S)-Methyl 1,2,3,4-tetrahydroisoquinoline-3-carboxylate hydrochloride 97%
(S)-Methyl 1,2,3,4-tetrahydroisoquinoline-3-carboxylate hydrochloride

Storage condition: Inert atmosphere, Room Temperature

Formel: C₁₁H₁₄ClNO₂
Molekylvægt: 227,6900 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD05861484
CAS-nummer: 78183-55-8

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR