4'-Methoxyacetoacetanilide 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD13205-10GEA 54.3 DKK
BLDPBD13205-10G BLDPBD13205-500G BLDPBD13205-100G BLDPBD13205-25G
4'-Methoxyacetoacetanilide 95%
4'-Methoxyacetoacetanilide

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₁H₁₃NO₃
Molekylvægt: 207,2300 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00008783
CAS-nummer: 5437-98-9

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR