N-(Cyanomethyl)-4-(trifluoromethyl)nicotinamide 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD130423-100GEA 458 DKK
BLDPBD130423-100G BLDPBD130423-5G BLDPBD130423-25G
N-(Cyanomethyl)-4-(trifluoromethyl)nicotinamide 95%
N-(Cyanomethyl)-4-(trifluoromethyl)nicotinamide

Storage condition: Sealed in dry, Room Temperature

Formel: C₉H₆F₃N₃O
Molekylvægt: 229,1587 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD07364026
CAS-nummer: 158062-67-0

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR