2-(4-(1-Hydroxy-4-(4-(hydroxydiphenylmethyl)piperidin-1-yl)butyl)phenyl)-2-methylpropanoic acid 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD130353-1GEA 551 DKK
BLDPBD130353-1G BLDPBD130353-250MG BLDPBD130353-5G
2-(4-(1-Hydroxy-4-(4-(hydroxydiphenylmethyl)piperidin-1-yl)butyl)phenyl)-2-methylpropanoic acid 95%
2-(4-(1-Hydroxy-4-(4-(hydroxydiphenylmethyl)piperidin-1-yl)butyl)phenyl)-2-methylpropanoic acid

Storage condition: Sealed in dry, Store in freezer, under -20°C

Formel: C₃₂H₃₉NO₄
Molekylvægt: 501,6564 g/mol
Opbevaringstemperatur: Fryser
MDL Number: MFCD00871892
CAS-nummer: 83799-24-0

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR