3-(Indol-3-yl)propionsyre 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD13027-500GEA 1590 DKK
BLDPBD13027-500G BLDPBD13027-25G BLDPBD13027-5G BLDPBD13027-10G BLDPBD13027-100G BLDPBD13027-1000G
3-(Indol-3-yl)propionsyre 97%
3-(Indol-3-yl)propionsyre

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₁H₁₁NO₂
Molekylvægt: 189,21 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00005660
CAS-nummer: 830-96-6

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR