2-Hydroxy-3a,4,7,7a-tetrahydro-1H-4,7-epoxyisoindole-1,3(2H)-dione 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD129954-250MGEA 179 DKK
BLDPBD129954-250MG
2-Hydroxy-3a,4,7,7a-tetrahydro-1H-4,7-epoxyisoindole-1,3(2H)-dione 95%
2-Hydroxy-3a,4,7,7a-tetrahydro-1H-4,7-epoxyisoindole-1,3(2H)-dione

Storage condition: Inert atmosphere, Store in freezer, under -20°C

Formel: C₈H₇NO₄
Molekylvægt: 181,14 g/mol
Opbevaringstemperatur: Fryser
MDL Number: MFCD00180244
CAS-nummer: 5596-17-8

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR