3-Propylidenephthalide cis- and trans-mixture 96%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD12963-25GEA 349 DKK
BLDPBD12963-0.025G BLDPBD12963-25G BLDPBD12963-1G BLDPBD12963-100G BLDPBD12963-5G
3-Propylidenephthalide cis- and trans-mixture 96%
3-Propylidenephthalide

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₁H₁₀O₂
Molekylvægt: 174,20 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00051813
CAS-nummer: 17369-59-4

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR