Ethyl β-Thioglucopyranoside 98+%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD129271-25GEA 3630 DKK
BLDPBD129271-25G BLDPBD129271-1G
Ethyl β-Thioglucopyranoside 98+%
Ethyl β-Thioglucopyranoside

Storage condition: Inert atmosphere, Store in freezer, under -20°C

Formel: C₈H₁₆O₅S
Molekylvægt: 224,27 g/mol
Opbevaringstemperatur: Fryser
MDL Number: MFCD09836206
CAS-nummer: 7473-36-1

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR