Ethyl acetopyruvate 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD129246-100GEA 791 DKK
BLDPBD129246-100G BLDPBD129246-10G BLDPBD129246-5G BLDPBD129246-25G
Ethyl acetopyruvate 98%
Ethyl acetopyruvate

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C₇H₁₀O₄
Molekylvægt: 158,15 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00009124
CAS-nummer: 615-79-2

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR