Ethyl 4-hydroxy-6-methyl-2-oxo-1,2-dihydropyridine-3-carboxylate 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD128899-100GEA 2780 DKK
BLDPBD128899-100G BLDPBD128899-5G BLDPBD128899-10G BLDPBD128899-25G
Ethyl 4-hydroxy-6-methyl-2-oxo-1,2-dihydropyridine-3-carboxylate 97%
Ethyl 4-hydroxy-6-methyl-2-oxo-1,2-dihydropyridine-3-carboxylate

Storage condition: Sealed in dry, Room Temperature

Formel: C₉H₁₁NO₄
Molekylvægt: 197,18 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00205625
CAS-nummer: 10350-10-4

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR