Ethyl 4-amino-2-(methylthio)thiazole-5-carboxylate 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD128813-25GEA 8120 DKK
BLDPBD128813-25G BLDPBD128813-250MG BLDPBD128813-1G BLDPBD128813-5G
Ethyl 4-amino-2-(methylthio)thiazole-5-carboxylate 95%
Ethyl 4-amino-2-(methylthio)thiazole-5-carboxylate

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formel: C₇H₁₀N₂O₂S₂
Molekylvægt: 218,3000 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD01830310
CAS-nummer: 39736-29-3

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR