Ethyl 2-methyl-1H-imidazole-4-carboxylate 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD128349-100MGEA 559 DKK
BLDPBD128349-100MG BLDPBD128349-1G BLDPBD128349-250MG
Ethyl 2-methyl-1H-imidazole-4-carboxylate 98%
Ethyl 2-methyl-1H-imidazole-4-carboxylate

Storage condition: Sealed in dry, Room Temperature

Formel: C₇H₁₀N₂O₂
Molekylvægt: 154,16 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD02646638
CAS-nummer: 87326-25-8

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR