Ethyl 2-(3,5-dimethoxyphenyl)acetate 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD127968-25GEA 3800 DKK
BLDPBD127968-25G BLDPBD127968-250MG BLDPBD127968-1G BLDPBD127968-5G BLDPBD127968-100MG BLDPBD127968-10G
Ethyl 2-(3,5-dimethoxyphenyl)acetate 98%
Ethyl 2-(3,5-dimethoxyphenyl)acetate

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₂H₁₆O₄
Molekylvægt: 224,25 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD11111879
CAS-nummer: 65976-77-4

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR