(R)-(-)-Ethyl mandelat 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD127771-100GEA 931 DKK
BLDPBD127771-100G BLDPBD127771-25G BLDPBD127771-5G
(R)-(-)-Ethyl mandelat 98%
(R)-(-)-Ethyl mandelat

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₀H₁₂O₃
Molekylvægt: 180,2 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00064248
CAS-nummer: 10606-72-1

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR