Dodecyl chloroformate 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD126782-25GEA 54.3 DKK
BLDPBD126782-25G
Dodecyl chloroformate 98%
Dodecyl chloroformate

Storage condition: Inert atmosphere, 2-8°C

Formel: C₁₃H₂₅ClO₂
Molekylvægt: 248,7894 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00059483
CAS-nummer: 24460-74-0

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR