D-(-)-Luciferin potassium salt 97%
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
|
BLDPBD126660-5GEA
5280
DKK
BLDPBD126660-5G
BLDPBD126660-250MG
BLDPBD126660-1G
BLDPBD126660-100MG
D-(-)-Luciferin potassium salt 97%
D-(-)-Luciferin potassium salt
Storage condition: Inert atmosphere, Store in freezer, under -20°C
Formel:
C₁₁H₇KN₂O₃S₂ Molekylvægt: 318,41 g/mol Opbevaringstemperatur: Fryser |
MDL Number:
MFCD00044928 CAS-nummer: 115144-35-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...