Dimethyl pyrimidine-4,6-dicarboxylate 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD125465-1GEA 838 DKK
BLDPBD125465-1G BLDPBD125465-250MG
Dimethyl pyrimidine-4,6-dicarboxylate 95%
Dimethyl pyrimidine-4,6-dicarboxylate

Storage condition: Sealed in dry, Room Temperature

Formel: C₈H₈N₂O₄
Molekylvægt: 196,16 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD05864412
CAS-nummer: 6345-43-3

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR