Furfuryl disulphide 98%
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
|
BLDPBD124977-10GEA
69.8
DKK
BLDPBD124977-10G
BLDPBD124977-25G
Furfuryl disulphide 98%
Furfuryl disulphide
Storage condition: Keep in dark place, Inert atmosphere, 2-8°C
Formel:
C₁₀H₁₀O₂S₂ Molekylvægt: 226,3152 g/mol Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD00010082 CAS-nummer: 4437-20-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...