Diethyl 2-(4-chlorophenyl)malonate 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD124621-1GEA 745 DKK
BLDPBD124621-1G BLDPBD124621-5G BLDPBD124621-250MG
Diethyl 2-(4-chlorophenyl)malonate 97%
Diethyl 2-(4-chlorophenyl)malonate

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₃H₁₅ClO₄
Molekylvægt: 270,70 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD06204602
CAS-nummer: 19677-37-3

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR