4-Cyano-4'-heptyloxybiphenyl 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD1243-5GEA 427 DKK
BLDPBD1243-5G BLDPBD1243-250MG BLDPBD1243-1G
4-Cyano-4'-heptyloxybiphenyl 97%
4-Cyano-4'-heptyloxybiphenyl

Storage condition: Sealed in dry, Room Temperature

Formel: C₂₀H₂₃NO
Molekylvægt: 293,41 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00134146
CAS-nummer: 52364-72-4

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR