Sodium-D(+)-glucuronate monohydrate 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD123963-100GEA 977 DKK
BLDPBD123963-100G BLDPBD123963-25G BLDPBD123963-5G BLDPBD123963-1G
Sodium-D(+)-glucuronate monohydrate 98%
Sodium-D(+)-glucuronate monohydrate

Storage condition: Inert atmosphere, Room Temperature

Formel: C₆H₁₀O₇·1H₂O
Molekylvægt: 234,14 g/mol (216,14 anhy)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00151051
CAS-nummer: 207300-70-7

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR