(3R,4S,5R)-3,4-Dihydroxy-5-((1S,2R)-1,2,3-trihydroxypropyl)dihydrofuran-2(3H)-one 95+%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD123938-100GEA 6700 DKK
BLDPBD123938-100G BLDPBD123938-25G BLDPBD123938-5G BLDPBD123938-1G
(3R,4S,5R)-3,4-Dihydroxy-5-((1S,2R)-1,2,3-trihydroxypropyl)dihydrofuran-2(3H)-one 95+%
(3R,4S,5R)-3,4-Dihydroxy-5-((1S,2R)-1,2,3-trihydroxypropyl)dihydrofuran-2(3H)-one

Storage condition: Sealed in dry, Room Temperature

Formel: C₇H₁₂O₇
Molekylvægt: 208,16 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD29039122
CAS-nummer: 221129-03-9

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR