Denatoniumbenzoat 99%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD123753-100GEA 559 DKK
BLDPBD123753-100G BLDPBD123753-5G BLDPBD123753-25G BLDPBD123753-500G BLDPBD123753-10G
Denatoniumbenzoat 99%
Denatoniumbenzoat

Storage condition: Inert atmosphere, Room Temperature

Formel: C₂₈H₃₄N₂O₃
Molekylvægt: 446,59 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00031578
CAS-nummer: 3734-33-6

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR