Copper Pyrophosphate 98% + (contains < 1% H₂O)

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD122186-25GEA 46.6 DKK
BLDPBD122186-25G BLDPBD122186-0.025
Copper Pyrophosphate 98% + (contains < 1% H₂O)
Copper Pyrophosphate

Storage condition: Inert atmosphere, Room Temperature

Formel: Cu₂O₇P₂
Molekylvægt: 301,03 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00016061
CAS-nummer: 10102-90-6

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR