N-(5-Chloro-4-((4-chlorophenyl)(cyano)methyl)-2-methylphenyl)-2-hydroxy-3,5-diiodobenzamide, sodium salt 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD122019-5GEA 101 DKK
BLDPBD122019-5G BLDPBD122019-500G BLDPBD122019-100G BLDPBD122019-25G BLDPBD122019-0.5G
N-(5-Chloro-4-((4-chlorophenyl)(cyano)methyl)-2-methylphenyl)-2-hydroxy-3,5-diiodobenzamide, sodium salt 98%
N-(5-Chloro-4-((4-chlorophenyl)(cyano)methyl)-2-methylphenyl)-2-hydroxy-3,5-diiodobenzamide, sodium salt

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₂₂H₁₄Cl₂I₂N₂NaO₂
Molekylvægt: 686,06 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD03427328
CAS-nummer: 61438-64-0

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR