(3,3-Diethoxyprop-1-en-1-yl)benzene 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD121350-100GEA 1090 DKK
BLDPBD121350-0.025 BLDPBD121350-100G BLDPBD121350-10G BLDPBD121350-25G BLDPBD121350-5G BLDPBD121350-1G
(3,3-Diethoxyprop-1-en-1-yl)benzene 95%
(3,3-Diethoxyprop-1-en-1-yl)benzene

Storage condition: Inert atmosphere, 2-8°C

Formel: C₁₃H₁₈O₂
Molekylvægt: 206,28 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00672803
CAS-nummer: 7148-78-9

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR