2-Octanamidoacetic acid 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD119870-500GEA 1590 DKK
BLDPBD119870-500G BLDPBD119870-25G BLDPBD119870-1000G BLDPBD119870-100G
2-Octanamidoacetic acid 97%
2-Octanamidoacetic acid

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₀H₁₉NO₃
Molekylvægt: 201,2600 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00154701
CAS-nummer: 14246-53-8

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR