Ethyl-α-bromophenylacetate 95%
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
BLDPBD11872-25GEA
1030
DKK
BLDPBD11872-25G
BLDPBD11872-10G
BLDPBD11872-5G
Ethyl-α-bromophenylacetate 95%
Ethyl-α-bromophenylacetate
Formel:
C₁₀H₁₁BrO₂ Molekylvægt: 243,10 g/mol |
MDL Number:
MFCD00013536 CAS-nummer: 2882-19-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...