6-Amino-2-methylquinoline 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD11778-5GEA 1790 DKK
BLDPBD11778-5G BLDPBD11778-250MG BLDPBD11778-1G
6-Amino-2-methylquinoline 98%
6-Amino-2-methylquinoline

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C₁₀H₁₀N₂
Molekylvægt: 158,20 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00052600
CAS-nummer: 65079-19-8

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR