Boc-ARG(Boc)2-OH 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD117766-1GEA 822 DKK
BLDPBD117766-1G BLDPBD117766-250MG BLDPBD117766-100MG BLDPBD117766-5G
Boc-ARG(Boc)2-OH 95%
Boc-ARG(Boc)2-OH

Storage condition: Sealed in dry, 2-8°C

Formel: C₂₁H₃₈N₄O₈
Molekylvægt: 474,555 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00270489
CAS-nummer: 97745-69-2

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR