12-((tert-Butoxycarbonyl)amino)dodecanoic acid 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD117664-250MGEA 148 DKK
BLDPBD117664-250MG BLDPBD117664-5G BLDPBD117664-25G BLDPBD117664-1G
12-((tert-Butoxycarbonyl)amino)dodecanoic acid 98%
12-((tert-Butoxycarbonyl)amino)dodecanoic acid

Storage condition: Sealed in dry, 2-8°C

Formel: C₁₇H₃₃NO₄
Molekylvægt: 315,44 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00235887
CAS-nummer: 18934-81-1

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR