(R)-3-(Boc-amino)-3-(3-chlorophenyl)propionic acid 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD117512-250MGEA 388 DKK
BLDPBD117512-250MG BLDPBD117512-100MG BLDPBD117512-1G
(R)-3-(Boc-amino)-3-(3-chlorophenyl)propionic acid 98%
(R)-3-(Boc-amino)-3-(3-chlorophenyl)propionic acid

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₁₄H₁₈BClNO₄
Molekylvægt: 299,75 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD03427959
CAS-nummer: 500789-06-0

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR