2-(1H-Pyrazol-1-yl)aniline 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD11690-250MGEA 140 DKK
BLDPBD11690-250MG BLDPBD11690-5G BLDPBD11690-1G
2-(1H-Pyrazol-1-yl)aniline 98%
2-(1H-Pyrazol-1-yl)aniline

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formel: C₉H₉N₃
Molekylvægt: 159,1879 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD03086227
CAS-nummer: 54705-91-8

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR