N-Benzyl-N-phenylaniline 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD116286-5GEA 140 DKK
BLDPBD116286-5G BLDPBD116286-25G
N-Benzyl-N-phenylaniline 95%
N-Benzyl-N-phenylaniline

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₉H₁₇N
Molekylvægt: 259,3450 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00092745
CAS-nummer: 606-87-1

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR