3-Hydroxy-4-methylbenzonitrile 97%
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
|
BLDPBD115862-100GEA
3280
DKK
BLDPBD115862-100G
BLDPBD115862-1G
BLDPBD115862-10G
BLDPBD115862-5G
BLDPBD115862-25G
3-Hydroxy-4-methylbenzonitrile 97%
3-Hydroxy-4-methylbenzonitrile
Storage condition: Sealed in dry, Room Temperature
Formel:
C₈H₇NO Molekylvægt: 133,1500 g/mol Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD08361769 CAS-nummer: 3816-66-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...