tert-Butyl benzoate 98%

Leverandør: BLD PHARMATECH GMBH
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD115787-100GEA 1030 DKK
BLDPBD115787-100G BLDPBD115787-5G BLDPBD115787-25G
tert-Butyl benzoate 98%
tert-Butyl benzoate

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₁H₁₄O₂
Molekylvægt: 178,2277 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00048244
CAS-nummer: 774-65-2

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR