(2S,5R)-Benzhydryl 3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate 4-oxide 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD115616-5GEA 652 DKK
BLDPBD115616-5G BLDPBD115616-1G BLDPBD115616-100G BLDPBD115616-25G
(2S,5R)-Benzhydryl 3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate 4-oxide 95%
(2S,5R)-Benzhydryl 3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate 4-oxide

Storage condition: Sealed in dry, 2-8°C

Formel: C₂₁H₂₁NO₄S
Molekylvægt: 383,4607 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD11044527
CAS-nummer: 87579-78-0

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR