BCAT 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD115232-5GEA 884 DKK
BLDPBD115232-5G BLDPBD115232-1G
BCAT 95%
BCAT

Storage condition: Sealed in dry, 2-8°C

Formel: C₁₄H₁₅N₅O₃S
Molekylvægt: 333,37 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD08274615
CAS-nummer: 163853-10-9

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR