Allyl a-D-glucopyranoside 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD113065-1GEA 1740 DKK
BLDPBD113065-1G BLDPBD113065-100MG BLDPBD113065-250MG
Allyl a-D-glucopyranoside 97%
Allyl a-D-glucopyranoside

Storage condition: Sealed in dry, 2-8°C

Formel: C₉H₁₆O₆
Molekylvægt: 220,21 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD04973540
CAS-nummer: 7464-56-4

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR