2,5-Dioxopyrrolidin-1-yl 6-acrylamidohexanoate 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD112614-1GEA 4170 DKK
BLDPBD112614-1G
2,5-Dioxopyrrolidin-1-yl 6-acrylamidohexanoate 97%
2,5-Dioxopyrrolidin-1-yl 6-acrylamidohexanoate

Storage condition: Inert atmosphere, Store in freezer, under -20°C

Formel: C₁₃H₁₈N₂O₅
Molekylvægt: 282,29 g/mol
Opbevaringstemperatur: Fryser
MDL Number: MFCD03427755
CAS-nummer: 63392-86-9

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR