2-Hydroxy-4-methylbenzamide 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD11201-5GEA 349 DKK
BLDPBD11201-5G BLDPBD11201-1G BLDPBD11201-25G
2-Hydroxy-4-methylbenzamide 95%
2-Hydroxy-4-methylbenzamide

Storage condition: Sealed in dry, Room Temperature

Formel: C₈H₉NO₂
Molekylvægt: 151,1626 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD02683864
CAS-nummer: 49667-22-3

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR