8-Isoquinolineboronic acid 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD111362-1GEA 838 DKK
BLDPBD111362-1G BLDPBD111362-100MG BLDPBD111362-5G BLDPBD111362-250MG
8-Isoquinolineboronic acid 97%
8-Isoquinolineboronic acid

Storage condition: Inert atmosphere, 2-8°C

Formel: C₉H₈BNO₂
Molekylvægt: 172,9800 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD11044913
CAS-nummer: 721401-43-0

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR