8-Fluoronaphthalen-2-ol 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD111277-100MGEA 737 DKK
BLDPBD111277-100MG
8-Fluoronaphthalen-2-ol 95%
8-Fluoronaphthalen-2-ol

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₀H₇FO
Molekylvægt: 162,1604 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD09037976
CAS-nummer: 13916-98-8

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR