8-Ethylquinoline 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD111267-1GEA 1430 DKK
BLDPBD111267-1G BLDPBD111267-250MG BLDPBD111267-100MG
8-Ethylquinoline 97%
8-Ethylquinoline

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₁H₁₁N
Molekylvægt: 157,2117 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00040688
CAS-nummer: 19655-56-2

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR