2-Methoxy-4-(trifluoromethyl)phenylboronic acid 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD11097-10GEA 1300 DKK
BLDPBD11097-10G BLDPBD11097-5G BLDPBD11097-1G BLDPBD11097-100G BLDPBD11097-100MG BLDPBD11097-25G BLDPBD11097-250MG
2-Methoxy-4-(trifluoromethyl)phenylboronic acid 98%
2-Methoxy-4-(trifluoromethyl)phenylboronic acid

Storage condition: Inert atmosphere, 2-8°C

Formel: C₈H₈BF₃O₃
Molekylvægt: 219,9500 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD03428511
CAS-nummer: 312936-89-3

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR