7-Fluoroquinazolin-4(3H)-one 95%
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
|
BLDPBD110613-10GEA
1010
DKK
BLDPBD110613-10G
BLDPBD110613-1G
BLDPBD110613-250MG
BLDPBD110613-100G
BLDPBD110613-5G
BLDPBD110613-25G
7-Fluoroquinazolin-4(3H)-one 95%
7-Fluoroquinazolin-4(3H)-one
Storage condition: Sealed in dry, Room Temperature
Formel:
C₈H₅FN₂O Molekylvægt: 164,1400 g/mol Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD07689443 CAS-nummer: 16499-57-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...